VWR® Bottles, Water Sampling
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
331-0074EA 708 DKK
331-0074 331-0076 VWRI331-0074.1 VWRI331-0075.1 VWRI331-0076.1 VWRI331-0077.1
VWR® Bottles, Water Sampling
Flasker Flasker til vandprøvetagning
PET, clear, with PP screw caps.
  • Dosed with sodium thiosulphate 18 mg (1,8%)
  • Capped, dosed and irradiated
  • Temperature resistance: –10 to +40 °C
  • Excellent shock resistance
  • Graduated
Ideal for water testing. Lightweight, rigid, durable, space saving bottles. Domed cap with tamper-evident security seal. Clear PET ensures good visibility of the content level.
Specific for UK demand.