Immobilisation kit for proteins, SulfoLink™, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20325 44995
PIER20325EA 2350 DKK
PIER20325 PIER44995
Immobilisation kit for proteins, SulfoLink™, Pierce™
Protein oprensnings Protein Purification Kits
SulfoLink™ Immobilisation Kit for Proteins is a kit containing all the reagents for preparing the protein sample for efficient immobilisation and columns containing SulfoLink™ resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification.
  • Specific to sulphhydryl (-SH) groups
  • Prepare and couple protein samples in three and half hours
  • Flexible coupling conditions, use pH 7,5 to 9,0 aqueous buffers, organic solvent or denaturant
  • Streamlined protocols for sample preparation, immobilisation, and affinity purification
  • Immobilise 2 to 20 mg of protein per 2 ml column of SulfoLink™ coupling resin
When incubated with a solution of protein that contains reduced cysteine residues, the iodoacetyl groups of SulfoLink™ resin react specifically and efficiently with the exposed sulphhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the protein to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.
Leveringsinformation: Standard kit includes SulfoLink™ columns, 2 ml (5 columns), SulfoLink™ preparation buffer (7,5 ml), SulfoLink™ coupling buffer (500 ml), wash solution (120 ml), PBS pack (makes 500 ml), 2-Mercaptoethylamine·HCl (5×6 mg), L-Cysteine·HCl (100 mg), Zeba™ spin desalting columns, 5 ml (5 columns), and column accessories. A 1-column trial kit is also available.