Cleanroom overalls with hood and integrated boots, BioClean-D™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S-BDFC-XL S-BDFC-L S-BDFC-XXL S-BDFC-XXXL S-BDFC-S S-BDFC-XS S-BDFC-M S-BDFC-XXXXL BDFC-L BDFC-XL BDFC-XXL
NITRS-BDFC-XLEA 4000 DKK
NITRS-BDFC-XL NITRS-BDFC-L NITRS-BDFC-XXL NITRS-BDFC-XXXL NITRS-BDFC-S NITRS-BDFC-XS NITRS-BDFC-M NITRS-BDFC-XXXXL NITRBDFC-L NITRBDFC-XL NITRBDFC-XXL
Cleanroom overalls with hood and integrated boots, BioClean-D™
Engangsbeklædning
The BioClean-D overall with hood and integrated boots is a disposable garment offering comfort and head-to-toe protection.
  • Light weight and low-linting
  • CleanTough material
  • Thumb loops to ensure a secure hold
  • Zip with sealable cover
  • Elasticated hood, back, cuffs and ankles
  • Slip-resistant soles
The BioClean-D overall with hood and integrated boots features a front zip closure with protective flap, elasticated hood, back, cuffs and ankles, and thumb loops to ensure a secure hold. The integrated boots feature slip-resistant soles to ensure every step is taken with confidence.
Certificeringer: EN ISO 13982-1:2004+A1:2009 (Type 5), EN ISO 13034:2005+A1:2010 (Type 6), EN 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile models: Sterilised in accordance with ISO 11137-1:2006.
Emballering: One piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 20 outer bags per lined carton (20 pieces). All sizes subject to minimum order quantity and lead time.