Protein A Plus agarose, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22811 22812 22810
PIER22811EA 5260 DKK
PIER22811 PIER22812 PIER22810
Protein A Plus agarose, Pierce™
Protein oprensnings Protein Purification Resins
Pierce™ Protein A Plus agarose is a high capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilised Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • High capacity beads with a dense load of immobilised Protein A
Binding capacity: >34 mg human IgG/ml resin (approx. 16 to 17 mg mouse IgG/ml resin)