Protein A/G Plus agarose, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20424 20423
PIER20424EA 28000 DKK
PIER20424 PIER20423
Protein A/G Plus agarose, Pierce™
Protein oprensnings Protein Purification Resins
Pierce™ protein A/G plus agarose is a high capacity agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilised protein A/G is ideal for polyclonal IgG purification from nearly any mammalian species
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A/G is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • High capacity beads with a dense load of immobilised Protein A
Binding capacity: 27 mg mouse IgG or ≥50 mg human IgG/ml of settled resin.