Test strips, peracetic acid, Reflectoquant®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.16975.0001 1.16976.0001 1.17956.0001
1.16975.0001EA 401 DKK
1.16975.0001 1.16976.0001 1.17956.0001
Test strips, peracetic acid, Reflectoquant®, Supelco®
Miljø test kit
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Applications: Disinfectant solutions - in cases in which hydrogen peroxide is present.