Affinity chromatography cartridges, cobalt, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
90093 90094
PIER90093EA 1960 DKK
PIER90093 PIER90094
Affinity chromatography cartridges, cobalt, Pierce™
Kromatografikolonner
Pierce™ HisPur™ cobalt chromatography cartridges are convenient, ready-to-use pre-packed devices for efficient purification of polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells. His-tagged proteins are purified from total soluble protein extract using a cobalt-charged tetradentate chelator immobilised onto 6% crosslinked agarose.
  • Binds at least 10 mg of purified 6×His-tagged protein per ml of resin
  • Cobalt-chelate coordination core binds fewer host protein contaminants, resulting in lower background than nickel resins
  • Low metal leaching means no metal contamination in eluted histidine-tagged protein sample
HisPur™ cobalt chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Leveringsinformation: Kit contains an accessory pack (1 female Luer-Lok® Adapter, 1 connector fitting, 1 column plug and 1 or 2 bottom caps).