Pierce™ Chemiluminescent Nucleic Acid Detection Module Kit
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
89880 89880A
PIER89880EA 5060 DKK
PIER89880 PIER89880A
Pierce™ Chemiluminescent Nucleic Acid Detection Module Kit
Elektroforese reagenser Western Blotting-reagenser
The Pierce™ Chemiluminescent nucleic acid detection module is a complete system for the detection of biotin-labeled nucleic acids for various blotting applications, including Northern/Southern blots, ribonuclease protection assays (RPAs), and electrophoretic mobility shift assays (EMSAs) or gel-shift assays.
  • Fast and sensitive
  • Versatile kit protocol (included) is applicable to any biotin-based nucleic acid detection system: Northerns, Southerns, EMSA, RPAs and colony lifts
  • Kit contains only the critical detection components for those who already have biotin-labeled probes and an optimised hybridisation method developed
The system uses an enhanced luminol substrate for horseradish peroxidase (HRP) with optimised blocking and wash steps that together produce sensitivity equivalent to radioactive (P³²) systems and provide a clean, consistent background required for publication-quality data.
Leveringsinformation: Kit contains stabilised Streptavidin-HRP conjugate, 1,5 ml; chemiluminescent substrate (luminol/enhancer solution, 80 ml and stable peroxide solution, 80 ml); blocking buffer, 500 ml; 4X wash buffer, 500 ml; and substrate equilibration buffer, 500 ml.