Handheld turbidity meters, Turb® 430
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
600320 600325 600326 600321
705-1027EA 21510 DKK
705-1027 662-2147 705-0672 705-0671
Handheld turbidity meters, Turb® 430
Turbiditetsmålere
The Turb 430, with lab option via LabStation, include drinking water testing, environmental monitoring, laboratory reference measurements and process control.
  • Nephelometric measurements at 90° scattered light
  • Optional sets include rugged carrying case for field use
  • LabStation LSFlex for easy lab use as optional accessories
Certificeringer: Scattered light characteristics according to the European Pharmacopoeia 5,0.
AQA for data evaluation by interval setting and GLP compliant data export to a PC with LS data software option.
Leveringsinformation: Instruments (only) are supplied with calibration set (0,02; 10; 1000 NTU), two cuvettes, cleaning tissues and four batteries. Sets also include field case with table insert, calibration set, two cuvettes, screw driver and LS data PC software with connecting cable.