Microplates, steptavidin coated, high sensitivity, 96-well, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
15520 15525
PIER15520EA 3740 DKK
PIER15520 PIER15525
Microplates, steptavidin coated, high sensitivity, 96-well, Pierce™
Mikroplader Immunologi mikroplader
PS, streptavidin coated high sensitivity plates maximise capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.
  • Sensitive detection of low concentrations (ng/ml) of biotinylated IgG
  • Ready to use and pre-blocked with Blocker™ BSA buffer
  • Detection range is 5 to 300 ng/ml (100 μl) of biotinylated IgG
  • Activation level is 250 μl
Clear plates for colorimetric ELISA and other assays. Black plates for fluorescent assays.