Amine reactive biotinylation kits, EZ-Link™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21445 21455 21435 21425 21925 21935 21955 21945
PIER21445EA 4110 DKK
PIER21445 PIER21455 PIER21435 PIER21425 PIER21925 PIER21935 PIER21955 PIER21945
Amine reactive biotinylation kits, EZ-Link™
Proteinkoblings- og mærkningssystemer
The amine group is the most common target for modifying protein molecules because it is abundant in the majority of proteins either due to the presence of lysine bearing amino side chain functionality or the N-terminal-amine. Amine-reactive biotinylation reagents can be divided into two groups based on water solubility: NHS-esters and sulfo-NHS-esters.
NHS-esters of biotin are water-insoluble and membrane-permeable because they do not possess a charged group. They may be used for biotinylating internal as well as external cellular components. Since NHS-esters of biotin are water-insoluble, for reactions in aqueous solution they must first be dissolved in an organic solvent, such as DMSO and DMF.
Sulfo-NHS-esters are water soluble and useful for applications that cannot tolerate organic solvents. Sulfo-NHS-esters of biotin are recommended for use as cell surface biotinylation reagents. Because of the charged sulfonate group, these compounds do not penetrate the plasma membrane, thus sulfo-NHS-esters biotinylation is restricted to the cell surface.
EZ-Link™ Sulfo-NHS Biotinylation Kits: Complete kit with an optimised protocol for labelling a protein and determining how much biotin has been attached. Fast and efficient labelling procedure.
EZ-Link™ Sulfo-NHS-LC Biotinylation Kits: Complete kit with an optimised protocol for labelling a protein and determining how much biotin has been attached. Fast and efficient labelling procedure.
EZ-Link™ NHS-PEG4 Biotinylation Kits: PEG4-Biotin transfers water-solubility to biotinylated molecules.
EZ-Link™ Sulfo-NHS-SS Biotinylation Kits: Cleavable biotinylation reagent allows for later removal of the biotin label.