Sporicide, Contec® Sterile Prochlor
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SBT102PC SBC502PC SBC102PC
141-1374EA 3590 DKK
141-1374 141-1373 CONTSBC102PC
Sporicide, Contec® Sterile Prochlor
Renseopløsninger Disinfektanter
Contec® ProChlor is a ready to use sporicidal biocide based on stabilised hypochlorous acid with purified water and is specifically designed for cleanroom environments.
  • Efficacious against bacteria, fungi, moulds, yeasts, viruses and spores
  • Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
  • Sterile version ideal for Grade A and B cleanrooms
  • Contains no quaternary ammonium or surfactant so it's very low residue
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
Contec Sterile ProChlor is 0.2 micron filtered, and sterile filled in a Grade A environment. The sterile product is filled into pre-irradiated components. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Bestillingsoplysninger: A non-sterile version is also available.
Leveringsinformation: Supplied as either trigger sprays for small areas or 1L and 5L capped containers for when larger volumes are required.
Emballering: Double bagged packed in linear tear packaging.
Advarsel: Use biocides safely. Always read the label and product information before use.
No hazard classification means only basic PPE required and no special disposal is required.