Amine reactive biotinylation reagents, EZ-Link™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21336 20217 21330 21343 21312 21362 21441 21442 21313 21363
PIER21336EA 2180 DKK
PIER21336 PIER20217 PIER21330 PIER21343 PIER21312 PIER21362 PIER21441 PIER21442 PIER21313 PIER21363
Amine reactive biotinylation reagents, EZ-Link™
Proteinkoblings- og mærkningssystemer
The amine group is the most common target for modifying protein molecules because it is abundant in the majority of proteins either due to the presence of lysine bearing amino side chain functionality or the N-terminal-amine. Amine-reactive biotinylation reagents can be divided into two groups based on water solubility: NHS-esters and sulfo-NHS-esters.
NHS-esters of biotin are water-insoluble and membrane-permeable because they do not possess a charged group. They may be used for biotinylating internal as well as external cellular components. Since NHS-esters of biotin are water-insoluble, for reactions in aqueous solution they must first be dissolved in an organic solvent, such as DMSO and DMF.
Sulfo-NHS-esters are water soluble and useful for applications that cannot tolerate organic solvents. Sulfo-NHS-esters of biotin are recommended for use as cell surface biotinylation reagents. Because of the charged sulfonate group, these compounds do not penetrate the plasma membrane, thus sulfo-NHS-esters biotinylation is restricted to the cell surface.
PEGylated esters transfer water solubility to the biotinylated molecule, thus reducing aggregation of labeled proteins stored in solution. The PEG spacer arm also gives the reagent a long and flexible connection to minimise steric hindrance for binding to avidin molecules.