ESD safety shoes, lace-up, uvex 2, 6502
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6502.2/35 6502.2/36 6502.2/37 6502.2/38 6502.2/39 6502.2/40 6502.2/41 6502.2/42 6502.2/43 6502.2/44 6502.2/45 6502.2/46 6502.2/47 6502.2/48 6502.2/49 6502.2/50 6502.2/51 6502.2/52 6502.3/48 6502.3/49 6502.3/35 6502.3/42 6502.3/43 6502.3/38 6502.3/39 6502.3/40 6502.3/50 6502.3/41 6502.3/46 6502.3/51 6502.3/47 6502.3/52 6502.3/44 6502.3/45 6502.3/37 6502.3/36 6502.4/35 6502.4/50 6502.4/51 6502.4/52 6502.4/44 6502.4/45 6502.4/46 6502.4/47 6502.4/38 6502.4/39 6502.4/40 6502.4/41 6502.4/42 6502.4/43 6502.4/36 6502.4/37 6502.4/48 6502.4/49 6502.1/35 6502.1/36 6502.1/37 6502.1/38 6502.1/39 6502.1/40 6502.1/41 6502.1/42 6502.1/43 6502.1/44 6502.1/45 6502.1/46 6502.1/47 6502.1/48 6502.1/49 6502.1/50 6502.1/51 6502.1/52
114-6745EA 886 DKK
114-6745 114-6746 114-6747 114-6748 114-6749 114-6750 114-6751 114-6752 114-6753 114-6754 114-6755 114-6756 114-6757 114-6758 114-6759 114-6760 114-6761 114-6762 114-8315 114-8316 114-8302 114-8309 114-8310 114-8305 114-8306 114-8307 114-8317 114-8308 114-8313 114-8318 114-8314 114-8319 114-8311 114-8312 114-8304 114-8303 UVEX6502.4/35 UVEX6502.4/50 UVEX6502.4/51 UVEX6502.4/52 UVEX6502.4/44 UVEX6502.4/45 UVEX6502.4/46 UVEX6502.4/47 UVEX6502.4/38 UVEX6502.4/39 UVEX6502.4/40 UVEX6502.4/41 UVEX6502.4/42 UVEX6502.4/43 UVEX6502.4/36 UVEX6502.4/37 UVEX6502.4/48 UVEX6502.4/49 UVEX6502.1/35 UVEX6502.1/36 UVEX6502.1/37 UVEX6502.1/38 UVEX6502.1/39 UVEX6502.1/40 UVEX6502.1/41 UVEX6502.1/42 UVEX6502.1/43 UVEX6502.1/44 UVEX6502.1/45 UVEX6502.1/46 UVEX6502.1/47 UVEX6502.1/48 UVEX6502.1/49 UVEX6502.1/50 UVEX6502.1/51 UVEX6502.1/52
ESD safety shoes, lace-up, uvex 2, 6502
Sko og støvler
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water repellent leather upper, soft padded tongue and collar, plus a dual-density PUR sole free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction eliminates pressure points
  • Lightweight two-layer sole with excellent shock absorption and outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuff cap for additional protection
  • Flexible, metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including the chemical, transport, logistics, energy and mechanical engineering industries.
Colour: Black/orange
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, SRC