VWR®, Blue light LED transilluminators
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
730-1467EA 15670 DKK
730-1467 730-1468
VWR®, Blue light LED transilluminators
Lysbord Blåt lys Transilluminatorer
Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. A blue light transilluminator is a ‘safe’ light source, in that the user is not exposed to harmful UV radiation, and samples are free from photonicking.