VWR®, UV-transilluminatorer
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
730-1461EA 9030 DKK
730-1461 730-1462 730-1463 730-1480 730-1481 730-1472 730-1473 730-1474 730-1475 730-1476 730-1477 730-1478 730-1479 730-1483 730-1482 730-0497
VWR®, UV-transilluminatorer
Lysbord UV Transilluminatorer
All VWR® transilluminators slide out for easy access to provide very even illumination. Those for use with the VWR® Imager system also have variable intensity settings.
  • Choice of filter sizes
  • Safety cut-off (when in darkroom)
  • Choice of different/multiple wavelengths available
  • Available in Benchtop version with protective lid