Xanthine sodium salt ≥98%, piller
Leverandør: MP BiomedicalsSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
The methylated xanthine compounds caffeine, theobromine, and theophylline and their derivatives are used in medicine for their bronchodilator effects.
UV Visible Absorbance λ max (0.01 N HCl)=265 ± 5nm
Formel:
C₅H₃N₄NaO₂ |
MDL Number:
MFCD00069699 CAS-nummer: 1196-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...