Horse Apoferritin (from Spleen), MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0210026080 0210026025
ICNA0210026080EA 2220 DKK
ICNA0210026080 ICNA0210026025
Horse Apoferritin (from Spleen), MP Biomedicals
Proteiner
Soluble in water or dilute buffer.
  • Protein Content: Approximately 90%
  • Iro content: Typically less than 0,01%
Apoferritin is the protein shell of ferritin. It is very abundant in beta-cells of the pancreas.
It is widely used for the calibration of gel filtration columns and sodium dodecyl sulphate (SDS)-polyacrylamide gel electrophoresis. It acts as an anti-oxidant.
When added to cultured endothelial cells apoferritin is taken up in a dose-responsive manner and appears to protect the cells from oxidant-mediated cytolysis.