Gel loading-spidser, ART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2155 2155P 3621
732-2226EA 712 DKK
732-2226 732-2229 732-2255
Gel loading-spidser, ART™
Pipettespidser
These tips are designed for loading vertical acrylamide or agarose gels.
  • Sterile tips are certified free from RNase, DNase, ATP and pyrogen
  • Available with or without filter for trapping potential contaminants
  • Manufactured according to the industry's highest quality standard
På tilbud lige nu
De valgte varer nedenfor er i øjeblikket på udsalg
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering IRECS; ISO 14001 (Certificate Registration No. EMS 752603)