Barrier tips, extended, ART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2140-HR 2079-HR 2080-HR 2160P-HR
732-1140EA 534 DKK
732-1140 732-2398 732-2400 732-2406
Barrier tips, extended, ART™
Pipettespidser
These barrier tips are designed to fit virtually any pipette and application. The self-sealing barrier provides 100% protection against aerosols and liquid contamination.
  • Transparent, sterile
  • Free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
  • Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks, ensuring safe and secure storage