Barrier tips, low retention, extended length, ART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2140-05-HR 2079-05-HR 2080-05-HR 2160P-05-HR
732-1179EA 0 DKK
732-1179 732-2397 732-2399 732-2405
Barrier tips, low retention, extended length, ART™
Pipettespidser
These barrier tips are designed to fit virtually any pipette and application. The self-sealing barrier provides 100% protection against aerosols and liquid contamination. Using advanced low-retention technology, the hydrophobic inner surface of these tips significantly reduces sample loss to adhesion and maximises sample recovery.
  • Transparent, sterile
  • Free from RNase, DNase, DNA, ATP, bioburden, endotoxin, pyrogens and PCR inhibitors
  • Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
  • Rack cover can
More Product Information
På tilbud lige nu
De valgte varer nedenfor er i øjeblikket på udsalg
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering IRECS; ISO 14001 (Certificate Registration No. EMS 752603)