Pipette tips, low retention, extended length, ART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3510-05 3151-05-HR 3152-05-HR 3511-05-HR 3512-05-HR 3541-05-HR 3542-05-HR 3791-05-HR 3792-05-HR
732-0321EA 166 DKK
732-0321 613-2454 613-2456 613-2466 613-2468 613-2478 613-2487 613-2781 613-2783
Pipette tips, low retention, extended length, ART™
Pipettespidser
Using advanced low-retention technology, the hydrophobic inner surface of these tips significantly reduces sample loss to adhesion and maximises sample recovery.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
The tips are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
Emballering: Tips are available in bulk packaging or in racks.