Copper(II) gluconate
Leverandør: Spectrum Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
C1334-2.5KG C1334-25KGBL
SPCMC1334-2.5KGEA 15560 DKK
SPCMC1334-2.5KG SPCMC1334-25KGBL
Copper(II) gluconate
Copper(II) gluconate
Formel: C₁₂H₂₂CuO₁₄
Molekylvægt: 453.84 g/mol
MDL Number: MFCD00075297
CAS-nummer: 527-09-3