VWR®, Pulverpakker til bestemmelse af total nitrogen, reagenser til fotometre
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
85618.610EA 1050 DKK
85618.610 85619.610
VWR®, Pulverpakker til bestemmelse af total nitrogen, reagenser til fotometre
Miljø test kit
VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners
RS = Powder Pack reagent set