Polyvinylpyrrolidon, M.W. 1 300 000 USP (K90)
Leverandør: Spectrum Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
P1416-12KG P1416-2.5KG P1416-50KGBL
SPCMP1416-12KGEA 40180 DKK
SPCMP1416-12KG SPCMP1416-2.5KG SPCMP1416-50KGBL
Polyvinylpyrrolidon, M.W. 1 300 000 USP (K90)
Polyvinylpyrrolidon
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Formel: (C₆H₉NO)n
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01076626
CAS-nummer: 9003-39-8