Trimethyl orthovalerate 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
L20377.14
L20377.22
L20377.14EA
1230
DKK
L20377.14
L20377.22
Trimethyl orthovalerate 98%
Trimethyl orthovalerate
Purity: 98%
CAS No.: 13820-09-2
Molecular Formula: C8H18O3
Formula Weight: 162.23
Flash Point: 41°(105°F)
Refractive Index: 1.4100
Boiling Point: 164-166°
Beilstein Reference: 1739200
UN No.: UN1993
MDL No.: MFCD00008481
Formel:
C₈H₁₈O₃ Kogepunkt: 164…166 °C Massefylde: 0,941 Flammepunkt: 41 °C (105 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008481 CAS-nummer: 13820-09-2 EINECS: 237-496-5 ADR: 3,III Merck Index: 14,9717 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...