3-Acetoxy-1-propenylboronic acid pinacol ester 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(E)-3-Acetoxy-1-propenylboronic acid pinacol ester, (E)-3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)allyl acetate
L19700.03
L19700.MD
L19700.03EA
3010
DKK
L19700.03
L19700.MD
3-Acetoxy-1-propenylboronic acid pinacol ester 97%
3-Acetoxy-1-propenylboronic acid pinacol ester
Purity: 97%
CAS No.: 161395-97-7
Molecular Formula: C11H19BO4
Formula Weight: 226.08
Refractive Index: 1.4485
Beilstein Reference: 6923397
MDL No.: MFCD03788751
Formel:
C₁₁H₁₉BO₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03788751 CAS-nummer: 161395-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...