6-(Benzyloxy)-1H-indole 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L17805.03 L17805.MD
L17805.03EA 1010 DKK
L17805.03 L17805.MD
6-(Benzyloxy)-1H-indole 97%
6-(Benzyloxy)-1H-indole

Purity: 97% CAS No.: 15903-94-3 Molecular Formula: C15H13NO Formula Weight: 223.28 Melting Point: 114-116° Beilstein Reference: 173319 MDL No.: MFCD00053554

Formel: C₁₅H₁₃NO
Smeltepunkt: 114…116 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00053554
CAS-nummer: 15903-94-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR