N-(4-Chloro-2-butynyl)phthalimide 97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 4819-69-6
Molecular Formula: C12H8ClNO2
Formula Weight: 233.66
Odour: Nearly Odourless
Melting Point: 115-120°
Beilstein Reference: 179626
UN No.: UN3261
MDL No.: MFCD01318120
Formel:
C₁₂H₈ClNO₂ Smeltepunkt: 115…120 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01318120 CAS-nummer: 4819-69-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...