4,4-Dimethyl-1-phenylpentane-1,3-dione 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L15251.06
L15251.14
L15251.06EA
795
DKK
L15251.06
L15251.14
4,4-Dimethyl-1-phenylpentane-1,3-dione 99%
4,4-Dimethyl-1-phenylpentane-1,3-dione
Purity: 99%
CAS No.: 13988-67-5
Molecular Formula: C13H16O2
Formula Weight: 204.27
Refractive Index: 1.5590
Boiling Point: 127-128°/2mm
Beilstein Reference: 1868436
MDL No.: MFCD00511275
Formel:
C₁₃H₁₆O₂ Kogepunkt: 127…128 °C (2 mmHg) Massefylde: 1,024 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00511275 CAS-nummer: 13988-67-5 |