Diethylmethoxyborane 1 M THF

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L15107.ACEA 649 DKK
L15107.AC L15107.AE
Diethylmethoxyborane 1 M THF
Diethylmethoxyborane

CAS No.: 7397-46-8 Molecular Formula: C5H13BO Formula Weight: 99.97 Flash Point: -6°(21°F) Refractive Index: 1.4040 Beilstein Reference: 2231225 UN No.: UN2924 MDL No.: MFCD00013240

Formel: C₅H₁₃BO
Molekylvægt: 99.968 g/mol
Massefylde: 0.868
Flammepunkt: –6 °C (21 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013240
CAS-nummer: 7397-46-8
UN nummer: 2924
ADR: 3 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR