Pentaethylene glycol di-p-toluenesulphonate 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L14369.03
L14369.06
L14369.03EA
380
DKK
L14369.03
L14369.06
Pentaethylene glycol di-p-toluenesulphonate 95%
Pentaethylene glycol di-p-toluenesulphonate
Purity: 95%
CAS No.: 41024-91-3
Molecular Formula: C24H34O10S2
Formula Weight: 546.66
Flash Point:
Formel:
C₂₄H₃₄O₁₀S₂ Massefylde: 1,260 Flammepunkt: >110 °C (230 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012204 CAS-nummer: 41024-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...