O-Benzylhydroxylamine 96%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 96%
CAS No.: 622-33-3
Molecular Formula: C7H9NO
Formula Weight: 123.15
Refractive Index: 1.5410
Boiling Point: 118-119°/30mm
Beilstein Reference: 1906691
UN No.: UN2735
MDL No.: MFCD00221709
Formel:
C₇H₉NO Kogepunkt: 118…119 °C (30 mmHg) Massefylde: 1,058 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00221709 CAS-nummer: 622-33-3 ADR: , |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...