Allyl vinyl ether 95% stabilised
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
0.1% Stabilised with potassium hydroxide
Stab. with 0.1% potassium hydroxide
Purity: 95%
CAS No.: 3917-15-5
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point:
Formel:
C₅H₈O Molekylvægt: 84.12 g/mol Kogepunkt: 65…68 °C Massefylde: 0.781 Flammepunkt: <–5 °C (23 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00039827 CAS-nummer: 3917-15-5 EINECS: 223-482-6 UN nummer: 3271 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...