Sodium hydrogen-L-tartrate, anhydrous 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
|
Synonymer:
Sodium hydrogen tartrate, Sodium bitartrate, L-Tartaric acid sodium salt
L12469.22
L12469.36
L12469.14
L12469.22EA
354
DKK
L12469.22
L12469.36
L12469.14
Sodium hydrogen-L-tartrate, anhydrous 98%
Sodium hydrogen-L-tartrate
Purity: 98%
CAS No.: 526-94-3
Molecular Formula: C4H5NaO6
Formula Weight: 172.07
Melting Point: ca 243° dec.
Beilstein Reference: 6099125
Optical Rotation: +24.5° (c=3 in water)
MDL No.: MFCD00065393
Formel:
C₄H₅NaO₆ Smeltepunkt: ∼ 243 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00065393 CAS-nummer: 526-94-3 EINECS: 208-400-9 Merck Index: 14,8589 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...