4-tert-Butylstyrene 94% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: stabiliseret med 4-tert-Butylpyrocatechol 50 ppm

L12356.AE L12356.AP
L12356.AEEA 385 DKK
L12356.AE L12356.AP
4-tert-Butylstyrene 94% stabilised
4-tert-Butylstyrene

Stab. with 50 ppm 4-tert-butylcatechol


Purity: 94% CAS No.: 1746-23-2 Molecular Formula: C12H16 Formula Weight: 160.26 Flash Point: 80°(176°F) Refractive Index: 1.5250 Melting Point: -38° Boiling Point: 219° Beilstein Reference: 1903714 MDL No.: MFCD00065126

Formel: C₁₂H₁₆
Kogepunkt: 219 °C
Smeltepunkt: –38 °C
Massefylde: 0,884
Flammepunkt: 80 °C (176 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00065126
CAS-nummer: 1746-23-2
EINECS: 217-126-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR