2-Methoxyethyl acrylate 98% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethylene glycol methyl ether acrylate, Ethylene Glycol Monomethyl Ether Acrylate, (2-Methoxyethyl) acrylate, Acrylic Acid 2-Methoxyethyl Ester

Stabilization: 50 ppm-100 ppm Stabilised with 4-methoxyphenol (MEHQ)

L11902.18 L11902.30 L11902.0B
L11902.18EA 261 DKK
L11902.18 L11902.30 L11902.0B
2-Methoxyethyl acrylate 98% stabilised
2-Methoxyethyl acrylate

Stab. with ca 50-100ppm 4-methoxyphenol


Purity: 98% CAS No.: 3121-61-7 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 56°(133°F) Refractive Index: 1.4280 Odour: Ester-like Melting Point: -30° Boiling Point: 162° Beilstein Reference: 1754333 UN No.: UN2929 MDL No.: MFCD00048149

Formel: C₆H₁₀O₃
Molekylvægt: 130.14 g/mol
Kogepunkt: 162 °C
Smeltepunkt: < –30 °C
Massefylde: 1.008
Flammepunkt: 56 °C (133 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00048149
CAS-nummer: 3121-61-7
EINECS: 221-499-3
UN nummer: 2929
ADR: 6.1 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR