4-Phenoxyphenyl isocyanate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L11686.06 L11686.03
L11686.06EA 1290 DKK
L11686.06 L11686.03
4-Phenoxyphenyl isocyanate 98%
4-Phenoxyphenyl isocyanate

Purity: 98% CAS No.: 59377-19-4 Molecular Formula: C13H9NO2 Formula Weight: 211.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5950 Boiling Point: 169-170°/3mm Beilstein Reference: 393196 UN No.: UN2206 MDL No.: MFCD00013876

Formel: C₁₃H₉NO₂
Kogepunkt: 169…170 °C (3 mmHg)
Massefylde: 1,169
Flammepunkt: >110 °C (230 °F)
MDL Number: MFCD00013876
CAS-nummer: 59377-19-4
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR