3-Hydroxyphenylacetylene 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Ethynylphenol

L11523.03 L11523.06
L11523.03EA 1120 DKK
L11523.03 L11523.06
3-Hydroxyphenylacetylene 97%
3-Hydroxyphenylacetylene

Purity: 97% CAS No.: 10401-11-3 Molecular Formula: C8H6O Formula Weight: 118.14 Refractive Index: 1.5870 Boiling Point: 231° Beilstein Reference: 2076613 MDL No.: MFCD00078347

Formel: C₈H₆O
Molekylvægt: 118.14 g/mol
Kogepunkt: 231 °C
Massefylde: 1.09
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00078347
CAS-nummer: 10401-11-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR