4-Chlorocinnamamide 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L11406.03 L11406.06
L11406.03EA 635 DKK
L11406.03 L11406.06
4-Chlorocinnamamide 97%
4-Chlorocinnamamide

Purity: 97% CAS No.: 18166-64-8 Molecular Formula: C9H8ClNO Formula Weight: 181.62 Melting Point: 210-212° Beilstein Reference: 2084774 MDL No.: MFCD00017147

Formel: C₉H₈ClNO
Smeltepunkt: 210…212 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00017147
CAS-nummer: 18166-64-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR