Benzyl methacrylate 98% stabilised
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methacrylic Acid Benzyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 50 ppm-100 ppm
Stab. with ca 50-100ppm 4-methoxyphenol
Purity: 98%
CAS No.: 2495-37-6
Molecular Formula: C11H12O2
Formula Weight: 176.22
Flash Point: 107°(225°F)
Refractive Index: 1.5140
Boiling Point: 231-233°
Beilstein Reference: 2045924
MDL No.: MFCD00053684
Formel:
C₁₁H₁₂O₂ Kogepunkt: 231…233 °C Massefylde: 1,040 Flammepunkt: 107 °C (225 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00053684 CAS-nummer: 2495-37-6 EINECS: 219-674-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...