4,4'-Dichlorochalcone ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L10717.06
L10717.14
L10717.06EA
327
DKK
L10717.06
L10717.14
4,4'-Dichlorochalcone ≥98%
4,4'-Dichlorochalcone
Purity: 98+%
CAS No.: 19672-59-4
Molecular Formula: C15H10Cl2O
Formula Weight: 277.15
Melting Point: 156-160°
Beilstein Reference: 2331589
MDL No.: MFCD00018704
Formel:
C₁₅H₁₀Cl₂O Molekylvægt: 277.15 g/mol Smeltepunkt: 156…160 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00018704 CAS-nummer: 19672-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...