(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Naproxen, (S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

L09855.03 L09855.06 L09855.14
L09855.03EA 303 DKK
L09855.03 L09855.06 L09855.14
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 99%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Purity: 99% CAS No.: 22204-53-1 Molecular Formula: C14H14O3 Formula Weight: 230.26 Odour: Odourless Melting Point: 154-156° Beilstein Reference: 3591067 Optical Rotation: +66° (c=1 in chloroform) UN No.: UN2811 MDL No.: MFCD00010500

Formel: C₁₄H₁₄O₃
Molekylvægt: 230.26 g/mol
Smeltepunkt: 154…156 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010500
CAS-nummer: 22204-53-1
EINECS: 244-838-7
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,6417

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR