n-Propyl acrylate 95% stabilised
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 0,1%
L09714.09
L09714.18
L09714.09EA
881
DKK
L09714.09
L09714.18
n-Propyl acrylate 95% stabilised
n-Propyl acrylate
Stab. with 0.1% 4-methoxyphenol
Purity: 95%
CAS No.: 925-60-0
Molecular Formula: CH2=CHCO2(CH2)2CH3
Formula Weight: 114.15
Flash Point: 24°(75°F)
Refractive Index: 1.4123
Boiling Point: 43-44°/40mm
Beilstein Reference: 1744954
UN No.: UN3272
MDL No.: MFCD00048693
Formel:
C₆H₁₀O₂ Kogepunkt: 43…44 °C (40 mmHg) Massefylde: 0,920 Flammepunkt: 24 °C (75 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00048693 CAS-nummer: 925-60-0 EINECS: 213-120-5 UN nummer: 3272 ADR: 3,III |