N-tert-Butylacetamide 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L08505.09 L08505.18
L08505.09EA 818 DKK
L08505.09 L08505.18
N-tert-Butylacetamide 98%
N-tert-Butylacetamide

Purity: 98% CAS No.: 762-84-5 Molecular Formula: CH3CONHC(CH3)3 Formula Weight: 115.18 Melting Point: 95-99° Beilstein Reference: 1699507 MDL No.: MFCD00038244

Formel: CH₃CONHC(CH₃)₃
Smeltepunkt: 95…99 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00038244
CAS-nummer: 762-84-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR