Carbonylcyanide-3-chlorophenylhydrazone 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: CCCP, Carbonyl cyanide m-chlorophenylhydrazone, Carbonyl cyanide m-chlorophenylhydrazone

L06932.MC L06932.ME
L06932.MCEA 394 DKK
L06932.MC L06932.ME
Carbonylcyanide-3-chlorophenylhydrazone 98%
Carbonylcyanide-3-chlorophenylhydrazone

Purity: 98% CAS No.: 555-60-2 Molecular Formula: C9H5ClN4 Formula Weight: 204.62 Storage Temperature: -30°C to -10°C Odour: Odourless Melting Point: ca 175° dec. Beilstein Reference: 1842102 UN No.: UN3439 MDL No.: MFCD00001848

Formel: C₉H₅ClN₄
Smeltepunkt: ∼ 175 °C (dec.)
MDL Number: MFCD00001848
CAS-nummer: 555-60-2
EINECS: 209-103-7
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR