m-Toluenesulphonic acid monohydrate 97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 312619-56-0
Molecular Formula: C7H8O3S.H2O
Formula Weight: 190.22 (172.20anhy)
Refractive Index: 1.5270
Boiling Point: 165°/0.1mm
Beilstein Reference: 2208421
UN No.: UN2586
MDL No.: MFCD00167130
Formel:
C₇H₈O₃S·H₂O Kogepunkt: 165 °C (0,1 mmHg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00167130 CAS-nummer: 312619-56-0 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...