Cholesterol pelargonate
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
CAS No.: 1182-66-7
Molecular Formula: C36H62O2
Formula Weight: 526.89
Odour: Odourless
Melting Point: 75-79°
Beilstein Reference: 3179820
Optical Rotation: -30° (c=5 in chloroform)
MDL No.: MFCD00003643
Formel:
C₃₆H₆₂O₂ Smeltepunkt: 75…79 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003643 CAS-nummer: 1182-66-7 EINECS: 214-658-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...