1-Methyl-1H-indole-3-carbaldehyde ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Methylindole-3-carboxaldehyde

L02212.03 L02212.06
L02212.03EA 431 DKK
L02212.03 L02212.06
1-Methyl-1H-indole-3-carbaldehyde ≥98%
1-Methyl-1H-indole-3-carbaldehyde

Purity: 98+% CAS No.: 19012-03-4 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 67-71° Beilstein Reference: 121302 MDL No.: MFCD00014570

Formel: C₁₀H₉NO
Smeltepunkt: 67…71 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014570
CAS-nummer: 19012-03-4
EINECS: 242-750-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR