2-Acetamidoacrylic acid 99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 99%
CAS No.: 5429-56-1
Molecular Formula: C5H7NO3
Formula Weight: 129.12
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 1812032
MDL No.: MFCD00004257
Formel:
C₅H₇NO₃ Smeltepunkt: ∼ 190 °C (dec.) |
MDL Number:
MFCD00004257 CAS-nummer: 5429-56-1 EINECS: 226-583-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...